Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Plcd1
Entrez Gene ID
18799
Gene Name
phospholipase C, delta 1
Gene Aliases
AW212592, C79986
Chromosome Location
Chr.9: 119071527 - 119093502 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001293648.1 NP_001280577.1 5 - 6 948 71
NM_019676.3 NP_062650.1 5 - 6 899 71
NR_121571.1 5 - 6 899 71
GenBank mRNA AF133125.1 - 5 - 6 881 71
AK028749.1 - 5 - 6 900 71
AK082890.1 - 5 - 6 891 71
BC015249.1 - 5 - 6 853 71
BC025798.1 - 5 - 6 875 71
JN635596.1 - 5 - 6 948 71
U85711.1 - 5 - 6 893 71

View Full Product Details