Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pde4b
Entrez Gene ID
18578
Gene Name
phosphodiesterase 4B, cAMP specific
Gene Aliases
Dpde4, R74983, dunce
Chromosome Location
Chr.4: 102087537 - 102607262 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001177980.1 NP_001171451.1 8 - 9 1733 80
NM_001177981.1 NP_001171452.1 8 - 9 1273 80
NM_001177982.1 NP_001171453.1 8 - 9 1009 80
NM_019840.2 NP_062814.2 13 - 14 1898 80
GenBank mRNA AF208023.2 - 13 - 14 1833 80
AF326555.1 - 13 - 14 1638 80
AF326556.1 - 13 - 14 1638 80
AJ297397.1 - 13 - 14 1833 80
AK051102.1 - 8 - 9 1274 80
AK150868.1 - 8 - 9 1238 80
AK161341.1 - 8 - 9 1010 80
BC007155.1 - 8 - 9 1080 80
BC023751.1 - 8 - 9 1709 80
BC071259.1 - 8 - 9 1249 80

View Full Product Details