The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019933.2 | NP_064317.2 | 3 - 4 | 430 | 70 |
GenBank mRNA | AF106702.1 | - | 2 - 3 | 258 | 70 |
AK046683.1 | - | 3 - 4 | 705 | 70 |