Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sertad2
Entrez Gene ID
58172
Gene Name
SERTA domain containing 2
Gene Aliases
AB041541, AU021878, Kiaa0127, MNCb-1504, SEI-2, Sei2, TRIP-Br2, mKIAA0127
Chromosome Location
Chr.11: 20542938 - 20653023 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021372.2 NP_067347.2 1 - 2 507 77
GenBank mRNA AB041541.1 - 1 - 2 426 77
BC014726.1 - 1 - 2 339 77

View Full Product Details