The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023217.4 | NP_075706.1 | 4 - 5 | 529 | 80 |
GenBank mRNA | AJ278829.1 | - | 4 - 5 | 461 | 80 |
AK003373.1 | - | 4 - 5 | 473 | 80 | |
AK012658.1 | - | 4 - 5 | 461 | 80 | |
AK047835.1 | - | 4 - 5 | 473 | 80 | |
AK049632.1 | - | 4 - 5 | 452 | 80 | |
BC043041.1 | - | 4 - 5 | 439 | 80 | |
BC051938.1 | - | 4 - 5 | 458 | 80 |