The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025358.3 | NP_079634.2 | 1 - 2 | 80 | 65 |
GenBank mRNA | AK003137.1 | - | 1 - 2 | 62 | 65 |
AK003214.1 | - | 1 - 2 | 53 | 65 | |
BC058378.1 | - | 1 - 2 | 48 | 65 |