The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025541.3 | NP_079817.1 | 2 - 3 | 450 | 78 |
GenBank mRNA | AK007804.1 | - | 2 - 3 | 306 | 78 |
AK010210.1 | - | 2 - 3 | 460 | 78 | |
AK012376.1 | - | 2 - 3 | 441 | 78 | |
AK031513.1 | - | 2 - 3 | 416 | 78 | |
AK088618.1 | - | 2 - 3 | 393 | 78 | |
BC027628.1 | - | 2 - 3 | 242 | 78 |