Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mterf3
Entrez Gene ID
66410
Gene Name
mitochondrial transcription termination factor 3
Gene Aliases
2410017I18Rik, Mterfd1
Chromosome Location
Chr.13: 66905456 - 66933072 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025547.3 NP_079823.2 3 - 4 630 75
GenBank mRNA AK002299.1 - 3 - 4 571 75
AK010525.1 - 3 - 4 625 75
AK011934.1 - 3 - 4 618 75
AK034645.1 - 3 - 4 597 75
BC025173.1 - 3 - 4 599 75
BC085282.1 - 3 - 4 600 75

View Full Product Details