The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001285435.1 | NP_001272364.1 | 2 - 3 | 157 | 68 |
NR_104339.1 | 2 - 3 | 157 | 68 | ||
NR_104340.1 | 2 - 3 | 157 | 68 | ||
GenBank mRNA | AK008355.1 | - | 2 - 3 | 122 | 68 |
AK008364.1 | - | 2 - 3 | 122 | 68 | |
AK008440.1 | - | 2 - 3 | 121 | 68 | |
AK029081.1 | - | 2 - 3 | 95 | 68 |