Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cap2
Entrez Gene ID
67252
Gene Name
CAP, adenylate cyclase-associated protein, 2 (yeast)
Gene Aliases
2810452G09Rik, AV045235
Chromosome Location
Chr.13: 46501903 - 46650281 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026056.4 NP_080332.1 5 - 6 761 63
GenBank mRNA AK010235.1 - 5 - 6 508 63
AK013331.1 - 5 - 6 518 63
AK014979.1 - 1 - 2 157 63
AK033071.1 - 5 - 6 517 63
AK035556.1 - 5 - 6 762 63
AK047701.1 - 3 - 4 328 63
AK076737.1 - 1 - 2 181 63
BC050752.1 - 5 - 6 522 63
BC057937.1 - 5 - 6 576 63

View Full Product Details