Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cers4
Entrez Gene ID
67260
Gene Name
ceramide synthase 4
Gene Aliases
2900019C14Rik, Lass4, Trh1
Chromosome Location
Chr.8: 4492987 - 4526308 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026058.4 NP_080334.3 7 - 8 795 75
GenBank mRNA AK013554.1 - 7 - 8 747 75
AK028937.1 - 7 - 8 721 75
AK028945.1 - 7 - 8 698 75
AK030280.1 - 7 - 8 735 75
AK134887.1 - 6 - 7 1454 75
AY029531.1 - 7 - 8 696 75
BC003946.1 - 7 - 8 682 75

View Full Product Details