The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001085448.1 | NP_001078917.1 | 3 - 4 | 696 | 84 |
NM_001085449.1 | NP_001078918.1 | 2 - 3 | 407 | 84 | |
NM_001085450.1 | NP_001078919.1 | 3 - 4 | 696 | 84 | |
NM_001085453.1 | NP_001078922.1 | 3 - 4 | 466 | 84 | |
NM_007615.4 | NP_031641.2 | 4 - 5 | 814 | 84 | |
GenBank mRNA | AB093237.1 | - | 4 - 5 | 530 | 84 |
AK133193.1 | - | 3 - 4 | 417 | 84 | |
AK161790.1 | - | 4 - 5 | 590 | 84 | |
AK164485.1 | - | 3 - 4 | 681 | 84 | |
AK168453.1 | - | 2 - 3 | 406 | 84 | |
AK195355.1 | - | 1 - 2 | 66 | 84 | |
BC043108.1 | - | 3 - 4 | 631 | 84 | |
BC046589.1 | - | 2 - 3 | 207 | 84 | |
BC054544.1 | - | 4 - 5 | 747 | 84 | |
Z17804.1 | - | - | 609 | 84 |