The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001085448.1 | NP_001078917.1 | 15 - 16 | 2978 | 126 |
NM_001085449.1 | NP_001078918.1 | 13 - 14 | 2671 | 126 | |
NM_001085450.1 | NP_001078919.1 | 15 - 16 | 2978 | 126 | |
NM_001085453.1 | NP_001078922.1 | 14 - 15 | 2730 | 126 | |
NM_007615.4 | NP_031641.2 | 16 - 17 | 3096 | 126 | |
GenBank mRNA | AB093237.1 | - | 16 - 17 | 2812 | 126 |
AK133193.1 | - | 15 - 16 | 2699 | 126 | |
AK161790.1 | - | 16 - 17 | 2872 | 126 | |
AK168453.1 | - | 13 - 14 | 2669 | 126 | |
BC043108.1 | - | 15 - 16 | 2913 | 126 | |
BC054544.1 | - | 16 - 17 | 3029 | 126 | |
Z17804.1 | - | - | 2873 | 126 |