The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007625.2 | NP_031651.2 | 3 - 4 | 402 | 70 |
GenBank mRNA | BC045162.1 | - | 3 - 4 | 386 | 70 |
BC117801.1 | - | 3 - 4 | 276 | 70 | |
U63387.1 | - | 3 - 4 | 314 | 70 |