The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164717.1 | NP_001158189.1 | 6 - 7 | 704 | 77 |
NM_008018.4 | NP_032044.2 | 6 - 7 | 704 | 77 | |
GenBank mRNA | AJ007012.1 | - | 6 - 7 | 704 | 77 |
BC118022.1 | - | 6 - 7 | 540 | 77 |