The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008023.1 | NP_032049.1 | 1 - 1 | 48 | 109 |
GenBank mRNA | X92591.1 | - | 1 - 1 | 48 | 109 |