The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008023.1 | NP_032049.1 | 1 - 1 | 975 | 97 |
GenBank mRNA | BC115976.1 | - | 2 - 2 | 665 | 97 |
BC139476.1 | - | 1 - 1 | 950 | 97 | |
BC139477.1 | - | 1 - 1 | 950 | 97 | |
X92591.1 | - | 1 - 1 | 975 | 97 |