Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
G0s2
Entrez Gene ID
14373
Gene Name
G0/G1 switch gene 2
Gene Aliases
AI255151, AV006465
Chromosome Location
Chr.1: 193272160 - 193273188 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008059.3 NP_032085.1 1 - 2 206 80
GenBank mRNA AK003165.1 - 1 - 2 128 80
BY124107.1 - 1 - 2 206 80
X95280.1 - 1 - 2 110 80

View Full Product Details