Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mep1a
Entrez Gene ID
17287
Gene Name
meprin 1 alpha
Gene Aliases
AI098089, AW107200, Mep-1, Mep-1a, Mep1
Chromosome Location
Chr.17: 43474324 - 43504144 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008585.2 NP_032611.2 6 - 7 433 94
GenBank mRNA AK093914.1 - 6 - 7 394 94
AK164734.1 - 6 - 7 388 94
BC015258.1 - 6 - 7 433 94
M74897.1 - 6 - 7 432 94

View Full Product Details