The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164167.1 | NP_001157639.1 | 5 - 6 | 912 | 117 |
NM_001164168.1 | NP_001157640.1 | 5 - 6 | 928 | 117 | |
NM_001164169.1 | NP_001157641.1 | 5 - 6 | 912 | 117 | |
NM_001164170.1 | NP_001157642.1 | 5 - 6 | 928 | 117 | |
NM_008602.4 | NP_032628.3 | 5 - 6 | 912 | 117 | |
GenBank mRNA | AF039567.1 | - | 4 - 5 | 357 | 117 |
AK014871.1 | - | 5 - 6 | 903 | 117 | |
AK029716.1 | - | 5 - 6 | 913 | 117 | |
AK078813.1 | - | 5 - 6 | 882 | 117 | |
AK086653.1 | - | 5 - 6 | 844 | 117 | |
BC005596.1 | - | 5 - 6 | 789 | 117 | |
BC034711.1 | - | 5 - 6 | 918 | 117 |