The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008618.3 | NP_032644.3 | 1 - 2 | 205 | 64 |
GenBank mRNA | AK002395.1 | - | 1 - 2 | 84 | 64 |
AK002404.1 | - | 1 - 2 | 64 | 64 | |
AK005237.1 | - | 1 - 2 | 100 | 64 | |
AK008270.1 | - | 1 - 2 | 77 | 64 | |
AK164785.1 | - | 1 - 2 | 86 | 64 | |
AK168545.1 | - | 1 - 2 | 43 | 64 | |
BC050940.1 | - | 1 - 2 | 71 | 64 | |
M29462.1 | - | 1 - 2 | 84 | 64 |