The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042760.1 | NP_001036225.1 | 5 - 6 | 751 | 103 |
NM_008767.2 | NP_032793.2 | 5 - 6 | 711 | 103 | |
GenBank mRNA | AB012084.1 | - | 5 - 6 | 751 | 103 |
AF028739.1 | - | 5 - 6 | 667 | 103 | |
AF037065.1 | - | 5 - 6 | 682 | 103 | |
BC003734.1 | - | 5 - 6 | 657 | 103 | |
BC015078.1 | - | 4 - 5 | 598 | 103 | |
BE865039.1 | - | 2 - 3 | 218 | 103 |