Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sat1
Entrez Gene ID
20229
Gene Name
spermidine/spermine N1-acetyl transferase 1
Gene Aliases
AA617398, SSAT, Sat
Chromosome Location
Chr.X: 155213126 - 155216449 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291865.1 NP_001278794.1 1 - 2 345 104
NM_009121.4 NP_033147.1 1 - 2 345 104
GenBank mRNA AK002531.1 - 1 - 2 246 104
AK154445.1 - 1 - 2 241 104
BC058696.1 - 1 - 2 305 104
BC108334.1 - 1 - 2 176 104
L10244.1 - 1 - 2 153 104

View Full Product Details