The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042613.1 | NP_001036078.1 | 2 - 3 | 270 | 65 |
NM_001042614.1 | NP_001036079.1 | 2 - 3 | 353 | 65 | |
NM_009155.3 | NP_033181.3 | 2 - 3 | 287 | 65 | |
GenBank mRNA | AI892015.1 | - | 2 - 3 | 362 | 65 |
AK002450.1 | - | 2 - 3 | 269 | 65 | |
AK077265.1 | - | 2 - 3 | 269 | 65 | |
AK146774.1 | - | 2 - 3 | 269 | 65 | |
AK148623.1 | - | 2 - 3 | 269 | 65 | |
AK159269.1 | - | 2 - 3 | 269 | 65 | |
AK159524.1 | - | 2 - 3 | 270 | 65 | |
AK159894.1 | - | 2 - 3 | 266 | 65 | |
AK159911.1 | - | 2 - 3 | 270 | 65 | |
AK159935.1 | - | 2 - 3 | 269 | 65 | |
AK167611.1 | - | 2 - 3 | 269 | 65 | |
AK168463.1 | - | 2 - 3 | 266 | 65 | |
AK168583.1 | - | 2 - 3 | 266 | 65 | |
AK168631.1 | - | 2 - 3 | 269 | 65 | |
AK168747.1 | - | 2 - 3 | 269 | 65 | |
AK168926.1 | - | 2 - 3 | 269 | 65 | |
AK169011.1 | - | 2 - 3 | 268 | 65 | |
AK169158.1 | - | 2 - 3 | 268 | 65 | |
AK169362.1 | - | 2 - 3 | 269 | 65 | |
AK190911.1 | - | 1 - 2 | 104 | 65 | |
AK196001.1 | - | 1 - 2 | 104 | 65 | |
AK201752.1 | - | 1 - 2 | 104 | 65 | |
AK203751.1 | - | 1 - 2 | 104 | 65 | |
BC001991.1 | - | 2 - 3 | 270 | 65 | |
CB231357.1 | - | 2 - 3 | 290 | 65 | |
CK627189.1 | - | 2 - 3 | 270 | 65 | |
X99807.1 | - | 1 - 2 | 330 | 65 |