Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Snta1
Entrez Gene ID
20648
Gene Name
syntrophin, acidic 1
Gene Aliases
AW228934, Snt1
Chromosome Location
Chr.2: 154376314 - 154408084 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009228.2 NP_033254.2 1 - 2 404 78
GenBank mRNA AK037183.1 - 1 - 2 302 78
AK137379.1 - 1 - 2 405 78
BB625367.1 - 1 - 2 397 78
BC018546.1 - 1 - 2 354 78
U00677.1 - 1 - 2 401 78

View Full Product Details