Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Coro1a
Entrez Gene ID
12721
Gene Name
coronin, actin binding protein 1A
Gene Aliases
Clabp, Lmb3, TACO, p57
Chromosome Location
Chr.7: 126699774 - 126704816 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001301374.1 NP_001288303.1 5 - 6 883 64
NM_009898.3 NP_034028.1 5 - 6 796 64
GenBank mRNA AF047388.1 - 4 - 5 630 64
AF143955.1 - 5 - 6 767 64
AF495468.1 - 5 - 6 734 64
AK040885.1 - 5 - 6 740 64
AK146290.1 - 5 - 6 734 64
AK150962.1 - 5 - 6 738 64
AK151755.1 - 5 - 6 738 64
AK155527.1 - 5 - 6 739 64
AK155860.1 - 5 - 6 734 64
AK165296.1 - 5 - 6 738 64
AK172447.1 - 5 - 6 734 64
BC002136.1 - 5 - 6 724 64
BC053398.1 - 5 - 6 714 64
U89399.1 - 1 - 2 166 64

View Full Product Details