Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cyp46a1
Entrez Gene ID
13116
Gene Name
cytochrome P450, family 46, subfamily a, polypeptide 1
Gene Aliases
Cyp46
Chromosome Location
Chr.12: 108334261 - 108362248 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010010.1 NP_034140.1 1 - 2 191 98
GenBank mRNA AF094479.1 - 1 - 2 191 98
AK140740.1 - 1 - 2 196 98
BC018307.1 - 1 - 2 214 98

View Full Product Details