Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cyp46a1
Entrez Gene ID
13116
Gene Name
cytochrome P450, family 46, subfamily a, polypeptide 1
Gene Aliases
Cyp46
Chromosome Location
Chr.12: 108334261 - 108362248 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010010.1 NP_034140.1 6 - 7 648 87
GenBank mRNA AF094479.1 - 6 - 7 648 87
AK140740.1 - 6 - 7 653 87
BC018307.1 - 6 - 7 671 87

View Full Product Details