The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010220.4 | NP_034350.1 | 7 - 8 | 972 | 77 |
GenBank mRNA | AK169795.1 | - | 7 - 8 | 913 | 77 |
BC015260.1 | - | 7 - 8 | 911 | 77 | |
CT010372.1 | - | 6 - 7 | 753 | 77 | |
U16959.1 | - | 7 - 8 | 972 | 77 | |
U36220.1 | - | 7 - 8 | 900 | 77 |