Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Smad2
Entrez Gene ID
17126
Gene Name
SMAD family member 2
Gene Aliases
7120426M23Rik, Madh2, Madr2, Smad-2, mMad2
Chromosome Location
Chr.18: 76241251 - 76311748 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252481.1 NP_001239410.1 4 - 5 964 112
NM_010754.5 NP_034884.2 4 - 5 853 112
GenBank mRNA AF005743.1 - 3 - 4 581 112
AK007817.1 - 4 - 5 630 112
AY334552.1 - 2 - 3 432 112
BC021342.1 - 4 - 5 652 112
BC089184.1 - 4 - 5 964 112
U60530.1 - 3 - 4 581 112

View Full Product Details