The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177950.1 | NP_001171421.1 | 8 - 9 | 1155 | 67 |
NM_001177951.1 | NP_001171422.1 | 8 - 9 | 1155 | 67 | |
NM_001177952.1 | NP_001171423.1 | 8 - 9 | 1149 | 67 | |
NM_001177953.1 | NP_001171424.1 | 8 - 9 | 1155 | 67 | |
NM_011285.2 | NP_035415.1 | 8 - 9 | 1155 | 67 | |
GenBank mRNA | AF044677.1 | - | 8 - 9 | 1073 | 67 |
AJ238396.1 | - | 8 - 9 | 1067 | 67 | |
AK017192.1 | - | 8 - 9 | 1087 | 67 | |
AK029864.1 | - | 8 - 9 | 1281 | 67 | |
AK038652.1 | - | 8 - 9 | 1119 | 67 | |
AK136298.1 | - | 8 - 9 | 1086 | 67 | |
AK138977.1 | - | 8 - 9 | 1150 | 67 | |
BC012661.1 | - | 8 - 9 | 1094 | 67 |