The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011302.3 | NP_035432.3 | 2 - 3 | 252 | 71 |
GenBank mRNA | AF073780.1 | - | 2 - 3 | 115 | 71 |
AF084561.1 | - | 2 - 3 | 123 | 71 | |
AJ011381.1 | - | 2 - 3 | 103 | 71 | |
BC046422.1 | - | 2 - 3 | 252 | 71 |