The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011302.3 | NP_035432.3 | 5 - 6 | 690 | 83 |
GenBank mRNA | AF073780.1 | - | 5 - 6 | 553 | 83 |
AF084561.1 | - | 5 - 6 | 561 | 83 | |
AJ011381.1 | - | 5 - 6 | 541 | 83 | |
BC046422.1 | - | 5 - 6 | 690 | 83 |