The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011359.2 | NP_035489.2 | 3 - 4 | 358 | 64 |
GenBank mRNA | BC061137.1 | - | 3 - 4 | 353 | 64 |
CJ093163.1 | - | 3 - 4 | 354 | 64 |