The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011842.3 | NP_035972.3 | 5 - 6 | 811 | 91 |
GenBank mRNA | AF159259.1 | - | 5 - 6 | 686 | 91 |
AK077548.1 | - | 5 - 6 | 793 | 91 | |
AK088158.1 | - | 5 - 6 | 724 | 91 | |
AK147099.1 | - | 5 - 6 | 790 | 91 | |
AK149834.1 | - | 5 - 6 | 857 | 91 | |
AK157717.1 | - | 5 - 6 | 715 | 91 | |
AK159979.1 | - | 5 - 6 | 787 | 91 | |
AK180159.1 | - | 3 - 4 | 238 | 91 | |
AK206505.1 | - | 2 - 3 | 208 | 91 | |
BC079847.1 | - | 5 - 6 | 811 | 91 |