The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011842.3 | NP_035972.3 | 12 - 13 | 1551 | 65 |
GenBank mRNA | AF159259.1 | - | 12 - 13 | 1426 | 65 |
AK077548.1 | - | 12 - 13 | 1533 | 65 | |
AK088158.1 | - | 12 - 13 | 1464 | 65 | |
AK147099.1 | - | 12 - 13 | 1530 | 65 | |
AK149834.1 | - | 12 - 13 | 1597 | 65 | |
AK159979.1 | - | 12 - 13 | 1527 | 65 | |
AK211323.1 | - | 2 - 3 | 174 | 65 | |
AK213860.1 | - | 3 - 4 | 179 | 65 | |
AK218441.1 | - | 2 - 3 | 174 | 65 | |
AK218740.1 | - | 2 - 3 | 174 | 65 | |
BC079847.1 | - | 12 - 13 | 1551 | 65 |