Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rngtt
Entrez Gene ID
24018
Gene Name
RNA guanylyltransferase and 5'-phosphatase
Gene Aliases
AU020997, HCE, MCE1
Chromosome Location
Chr.4: 33310297 - 33502610 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF025653.1 - 9 - 10 1058 83
AF034568.1 - 9 - 10 1112 83
AK002922.1 - 9 - 10 1161 83
AK142016.1 - 9 - 10 1155 83
AK150497.1 - 9 - 10 1165 83
AK151463.1 - 9 - 10 1179 83
AK160007.1 - 9 - 10 1167 83
AK160706.1 - 9 - 10 1145 83
AK169305.1 - 9 - 10 1131 83
BC043657.1 - 9 - 10 1130 83

View Full Product Details