Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rcn2
Entrez Gene ID
26611
Gene Name
reticulocalbin 2
Gene Aliases
AA408742, Tcbp49
Chromosome Location
Chr.9: 56041845 - 56059083 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011992.2 NP_036122.2 3 - 4 662 96
GenBank mRNA AF049125.1 - 3 - 4 651 96
AK041996.1 - - 659 96
AK076153.1 - 3 - 4 489 96
AK077486.1 - 3 - 4 638 96
AK078106.1 - 3 - 4 630 96
AK169765.1 - 3 - 4 636 96
BC132320.1 - 3 - 4 627 96
BC145668.1 - 3 - 4 627 96

View Full Product Details