The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001083959.1 | NP_001077428.1 | 4 - 5 | 486 | 85 |
NM_001083960.1 | NP_001077429.1 | 4 - 5 | 447 | 85 | |
NM_012046.2 | NP_036176.1 | 5 - 6 | 561 | 85 | |
GenBank mRNA | AF126400.1 | - | 4 - 5 | 486 | 85 |
AF149309.1 | - | 4 - 5 | 410 | 85 | |
AF163053.1 | - | 4 - 5 | 447 | 85 | |
AF163054.1 | - | 4 - 5 | 447 | 85 | |
AF165313.1 | - | 4 - 5 | 440 | 85 | |
AF167439.1 | - | 4 - 5 | 479 | 85 | |
AF169386.1 | - | 5 - 6 | 558 | 85 | |
AF173848.1 | - | 5 - 6 | 554 | 85 |