The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013715.2 | NP_038743.1 | 1 - 2 | 486 | 72 |
GenBank mRNA | AF065386.1 | - | 1 - 2 | 239 | 72 |
AF068223.1 | - | 1 - 2 | 254 | 72 | |
AK002761.1 | - | 1 - 2 | 268 | 72 | |
AK005728.1 | - | 1 - 2 | 273 | 72 | |
AK012499.1 | - | 1 - 2 | 273 | 72 | |
AK133073.1 | - | 1 - 2 | 266 | 72 | |
AK146271.1 | - | 1 - 2 | 273 | 72 | |
AK151492.1 | - | 1 - 2 | 272 | 72 | |
BC046753.1 | - | 1 - 2 | 232 | 72 | |
CX240457.1 | - | 1 - 2 | 454 | 72 | |
U70736.1 | - | 1 - 2 | 262 | 72 |