The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290717.1 | NP_001277646.1 | 5 - 6 | 753 | 76 |
NM_001290718.1 | NP_001277647.1 | 5 - 6 | 726 | 76 | |
NM_001290719.1 | NP_001277648.1 | 5 - 6 | 753 | 76 | |
NM_013757.2 | NP_038785.1 | 5 - 6 | 753 | 76 | |
GenBank mRNA | AB025258.1 | - | 5 - 6 | 724 | 76 |
AB025259.1 | - | 5 - 6 | 724 | 76 | |
AB120510.1 | - | 4 - 5 | 539 | 76 | |
AK030401.1 | - | 5 - 6 | 600 | 76 | |
AK161582.1 | - | 5 - 6 | 720 | 76 | |
BC026819.1 | - | 5 - 6 | 753 | 76 |