Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fbxw2
Entrez Gene ID
30050
Gene Name
F-box and WD-40 domain protein 2
Gene Aliases
2700071L08Rik, FBW2, Fwd2, MD6
Chromosome Location
Chr.2: 34804364 - 34826273 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164768.1 NP_001158240.1 2 - 3 820 84
NM_001164769.1 NP_001158241.1 4 - 5 879 84
NM_001164770.1 NP_001158242.1 4 - 5 787 84
NM_013890.4 NP_038918.3 4 - 5 879 84
GenBank mRNA AF140683.1 - 2 - 3 824 84
AK040764.1 - 4 - 5 846 84
AK078815.1 - 4 - 5 787 84
AK088898.1 - 2 - 3 559 84
AK179719.1 - 2 - 3 373 84
X54352.1 - 3 - 4 809 84

View Full Product Details