Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fbxw5
Entrez Gene ID
30839
Gene Name
F-box and WD-40 domain protein 5
Gene Aliases
AI159739, Fbw5
Chromosome Location
Chr.2: 25500767 - 25505471 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013908.4 NP_038936.1 1 - 2 112 99
GenBank mRNA AF176520.1 - 1 - 2 73 99
AK019715.2 - 1 - 2 113 99

View Full Product Details