The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016803.3 | NP_058083.2 | 3 - 4 | 755 | 98 |
GenBank mRNA | AB008937.1 | - | 2 - 3 | 374 | 98 |
BC055055.1 | - | 1 - 2 | 634 | 98 |