Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Runx3
Entrez Gene ID
12399
Gene Name
runt related transcription factor 3
Gene Aliases
AML2, Cbfa3, Pebp2a3
Chromosome Location
Chr.4: 135120645 - 135177990 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019732.2 NP_062706.2 4 - 5 1049 73
GenBank mRNA AF085246.1 - 2 - 3 285 73
AF155880.1 - 3 - 4 822 73
AK155436.1 - 4 - 5 1043 73
AK155802.1 - 4 - 5 1041 73

View Full Product Details