The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019735.4 | NP_062709.3 | 4 - 5 | 430 | 77 |
GenBank mRNA | AB028863.1 | - | 4 - 5 | 322 | 77 |
AK077705.1 | - | 4 - 5 | 431 | 77 | |
AK145450.1 | - | 4 - 5 | 432 | 77 | |
BC028434.1 | - | 4 - 5 | 373 | 77 |