The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019779.3 | NP_062753.3 | 8 - 9 | 1449 | 76 |
GenBank mRNA | AF195119.1 | - | 8 - 9 | 1449 | 76 |
BC068264.1 | - | 8 - 9 | 1468 | 76 |