The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020010.2 | NP_064394.2 | 5 - 6 | 1145 | 78 |
GenBank mRNA | AF166266.1 | - | 5 - 6 | 897 | 78 |
AK028355.1 | - | 5 - 6 | 838 | 78 | |
AK028815.1 | - | 5 - 6 | 902 | 78 | |
AK031059.1 | - | 5 - 6 | 878 | 78 | |
AK076983.1 | - | 5 - 6 | 878 | 78 | |
AK217526.1 | - | 1 - 2 | 189 | 78 | |
BC031813.1 | - | 5 - 6 | 872 | 78 |