The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164096.1 | NP_001157568.1 | 5 - 6 | 491 | 64 |
NM_025298.3 | NP_079574.2 | 6 - 7 | 569 | 64 | |
GenBank mRNA | AK005034.1 | - | 5 - 6 | 492 | 64 |
AK012184.1 | - | 6 - 7 | 529 | 64 | |
AK144721.1 | - | 6 - 7 | 531 | 64 | |
AK159723.1 | - | 6 - 7 | 526 | 64 | |
BC037637.1 | - | 6 - 7 | 519 | 64 |