The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190473.1 | NP_001177402.1 | 7 - 8 | 865 | 62 |
NM_001190474.1 | NP_001177403.2 | 7 - 8 | 917 | 62 | |
NM_007828.2 | NP_031854.1 | 6 - 7 | 885 | 62 | |
GenBank mRNA | AB007143.1 | - | 6 - 7 | 798 | 62 |
AK035171.1 | - | 5 - 6 | 487 | 62 | |
BC012210.1 | - | 7 - 8 | 883 | 62 | |
BC023238.1 | - | 6 - 7 | 937 | 62 | |
BC125443.1 | - | 6 - 7 | 885 | 62 | |
BC137680.1 | - | 6 - 7 | 885 | 62 |